Titanium(IV) oxide, anatase, 99.6% (metals basis)
Titanium(IV) oxide, anatase, 99.6% (metals basis)
Titanium(IV) oxide, anatase, 99.6% (metals basis)
Titanium(IV) oxide, anatase, 99.6% (metals basis)
Thermo Scientific Chemicals
Titanium(IV) oxide, anatase, 99.6% (metals basis)
CAS: 1317-70-0 | O2Ti | 79.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF036199.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTitanium(IV) oxide
Name NoteAnatase
CAS1317-70-0
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H351
Suspected of causing cancer.
View more
Titanium(IV) oxide is used as UV-resistant cloth, skin protecting cream, face washing milk, cream milk, powder make-up. It is also used as photocatalyst, self-cleaning glass, self-cleaning ceramics, antibacterial material, air purification, sewage handling, chemical industry. It is widely utilized in cosmetics, sunscreen cream, natural white moisture protection cream, beauty and whitening cream, dawn and night cream, moistening refresher, vanishing cream, chemical fiber, plastics, printing ink coating. It is likewise utilized for improving the impressionability and opacity of the paper and used for producing titanium, ferrotitanium alloy, carbide alloy etc. in the metallurgical industry.