Titanium foil, 0.05mm (0.002in) thick, annealed, 99.6% (metals basis)
Titanium foil, 0.05mm (0.002in) thick, annealed, 99.6% (metals basis)
Titanium foil, 0.05mm (0.002in) thick, annealed, 99.6% (metals basis)
Titanium foil, 0.05mm (0.002in) thick, annealed, 99.6% (metals basis)
Thermo Scientific Chemicals
Titanium foil, 0.05mm (0.002in) thick, annealed, 99.6% (metals basis)
Form: approx. 0.14g/25x25mm; Temper: soft | CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Catalog NumberQuantity
ALF035811.GH100 x 100 mm
Catalog number ALF035811.GH
Price (MYR)
1,458.00
EA
Add to cart
Quantity:
100 x 100 mm
Request bulk or custom format
Price (MYR)
1,458.00
EA
Add to cart
Specifications
Chemical Name or MaterialTitanium
ColorSilver to Gray
Name NoteAnnealed
CAS7440-32-6
AppearanceSilver Gray foil