Titanium(IV) isopropoxide, 99.995% (metals basis)
Titanium(IV) isopropoxide, 99.995% (metals basis)
Titanium(IV) isopropoxide, 99.995% (metals basis)
Titanium(IV) isopropoxide, 99.995% (metals basis)
Thermo Scientific Chemicals
Titanium(IV) isopropoxide, 99.995% (metals basis)
CAS: 546-68-9 | C12H28O4Ti | 284.219 g/mol
Have Questions?
Catalog NumberQuantity
ALF035794.1425 g
Catalog number ALF035794.14
Price (MYR)
1,413.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,413.00
EA
Add to cart
Specifications
Chemical Name or MaterialTitanium(IV) isopropoxide
CAS546-68-9
Health Hazard 1H226-H319-H331-H335-H336
Health Hazard 2GHS H Statement
H226-H319
Flammable liquid and vapor.
Causes serious eye irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P311-P370+P378q-P501c
View more
Titanium(IV) isopropoxide is used as a precursor for the preparation of titanium and barium-strontium-titanate thin films. It is useful to make porous titanosilicates and potential ion-exchange materials for cleanup of radioactive wastes. It is an active component of Sharpless epoxidation as well as involved in the synthesis of chiral epoxides. In Kulinkovich reaction, it is involved as a catalyst in the preparation of cyclopropanes.