Yttrium(III) chloride, ultra dry, 99.99% (REO)
Yttrium(III) chloride, ultra dry, 99.99% (REO)
Yttrium(III) chloride, ultra dry, 99.99% (REO)
Yttrium(III) chloride, ultra dry, 99.99% (REO)
Thermo Scientific Chemicals
Yttrium(III) chloride, ultra dry, 99.99% (REO)
CAS: 10361-92-9
Have Questions?
Catalog NumberQuantity
ALF035743.0910 g
Catalog number ALF035743.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialYttrium(III) chloride
Melting Point721°C
Name NoteUltra dry
CAS10361-92-9
ColorWhite to Yellow
View more
Yttrium(III) chloride is used as a precursor for titanium-containing ceramics. It is used as an analytical reagent, as a catalyst and in superconductors. It is also used in the production of yttrium alkyl alkoxide complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only