Cerium(IV) sulfate, 0.1N Standardized Solution
Cerium(IV) sulfate, 0.1N Standardized Solution
Cerium(IV) sulfate, 0.1N Standardized Solution
Cerium(IV) sulfate, 0.1N Standardized Solution
Thermo Scientific Chemicals
Cerium(IV) sulfate, 0.1N Standardized Solution
CAS: 13590-82-4 | CeO8S2 | 332.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number ALF035650.K7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Specifications
Chemical Name or MaterialCerium(IV) sulfate
CAS13590-82-4
Health Hazard 1H314-H335-H350
Health Hazard 3P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
Recommended StorageAmbient temperatures
View more
Cerium(IV) sulfate is a redox indicator and is used in analytical chemistry for redox titrations. In ionized form, ceric ion acts as a strong oxidizer under acidic conditions. It is also used to prepare spherical-like nanoparticles CeO2, nanoparticles for NO reductions' reactions.