Sodium thiosulfate, 1.0N Standardized Solution
Sodium thiosulfate, 1.0N Standardized Solution
Sodium thiosulfate, 1.0N Standardized Solution
Sodium thiosulfate, 1.0N Standardized Solution
Thermo Scientific Chemicals
Sodium thiosulfate, 1.0N Standardized Solution
CAS: 7772-98-7 | Na2O3S2 | 158.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number ALF035647.K7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium thiosulfate
CAS7772-98-7
Recommended StorageAmbient temperatures
EINECS Number231-867-5
TSCAYes