Sodium hydroxide, 40% w/v aq. soln.
Sodium hydroxide, 40% w/v aq. soln.
Sodium hydroxide, 40% w/v aq. soln.
Sodium hydroxide, 40% w/v aq. soln.
Thermo Scientific Chemicals
Sodium hydroxide, 40% w/v aq. soln.
CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Catalog NumberQuantity
ALF035639.K74 L
Catalog number ALF035639.K7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium hydroxide
CAS1310-73-2
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
View more
Sodium hydroxide, 40% w/v aq. solution is used as a chemical reactant or neutralization agent in various industries like chemical, pharmaceutical, textile, pulp and paper, metal and food industries and soap and detergent manufacturing. It is also used in the manufacture of synthetic fabrics such as rayon and also for manufacturing of food products. It is utilized for pH adjustment and formation of in situ sodium methylate in bioethanol and biodiesel processing.