Sodium hydroxide, 0.05N Standardized Solution
Sodium hydroxide, 0.05N Standardized Solution
Sodium hydroxide, 0.05N Standardized Solution
Sodium hydroxide, 0.05N Standardized Solution
Thermo Scientific Chemicals
Sodium hydroxide, 0.05N Standardized Solution
CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Catalog NumberQuantity
ALF035623.K21 L
Catalog number ALF035623.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium hydroxide
Name NoteStandardized Solution
CAS1310-73-2
Recommended StorageAmbient temperatures
EINECS Number215-185-5