Hydrochloric acid, 10% v/v aq. soln.
Hydrochloric acid, 10% v/v aq. soln.
Hydrochloric acid, 10% v/v aq. soln.
Hydrochloric acid, 10% v/v aq. soln.
Thermo Scientific Chemicals
Hydrochloric acid, 10% v/v aq. soln.
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantity
ALF035611.K74 L
Catalog number ALF035611.K7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Specifications
AppearanceColorless to pale yellow liquid
Chemical Name or MaterialHydrochloric acid
ColorColorless to Yellow
CAS7647-01-0
Health Hazard 1H290-H315-H318
View more
Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing.