Silver nitrate, 0.1N Standardized Solution
Silver nitrate, 0.1N Standardized Solution
Silver nitrate, 0.1N Standardized Solution
Silver nitrate, 0.1N Standardized Solution
Thermo Scientific Chemicals
Silver nitrate, 0.1N Standardized Solution
CAS: 7761-88-8 | AgNO3 | 169.87 g/mol
Have Questions?
Catalog NumberQuantity
ALF035601.K21 L
Catalog number ALF035601.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialSilver nitrate
CAS7761-88-8
ColorColorless
Health Hazard 1H290-H315-H319-H400-H410: May be corrosive to metals. Causes skin irritation. Causes serious eye irritation. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
DOT InformationDOT Class: 9, Packaging Group: III
View more
Silver nitrate, 1.0N Standardized Solution is used for volumetric analysis in laboratories. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.