Potassium hydroxide, 1.0N Standardized Solution
Potassium hydroxide, 1.0N Standardized Solution
Potassium hydroxide, 1.0N Standardized Solution
Potassium hydroxide, 1.0N Standardized Solution
Thermo Scientific Chemicals
Potassium hydroxide, 1.0N Standardized Solution
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Catalog NumberQuantity
ALF035592.K21 L
Catalog number ALF035592.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium hydroxide
ColorColorless
Name NoteStandardized Solution
CAS1310-58-3
Health Hazard 1H290-H314-H335