Ammonium thiocyanate, 0.1N Standardized Solution
Ammonium thiocyanate, 0.1N Standardized Solution
Ammonium thiocyanate, 0.1N Standardized Solution
Ammonium thiocyanate, 0.1N Standardized Solution
Thermo Scientific Chemicals
Ammonium thiocyanate, 0.1N Standardized Solution
CAS: 7732-18-5 | NH4SCN
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number ALF035580.K7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium thiocyanate
CAS7732-18-5
Recommended StorageAmbient temperatures
EINECS Number217-175-6
TSCAYes
View more
Ammonium thiocyanate, 0.1N standardized solution is used for volumetric analysis in laboratories.