Ammonium molybdate (di), 4% w/v aq. soln.
Ammonium molybdate (di), 4% w/v aq. soln.
Ammonium molybdate (di), 4% w/v aq. soln.
Ammonium molybdate (di), 4% w/v aq. soln.
Thermo Scientific Chemicals
Ammonium molybdate (di), 4% w/v aq. soln.
CAS: 27546-07-2 | H2MoO4 | 161.96 g/mol
Have Questions?
Catalog NumberQuantity
ALF035578.AP500 mL
Catalog number ALF035578.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium molybdate (di)
CAS27546-07-2
Recommended StorageAmbient temperatures
TSCAYes
Molecular FormulaH2MoO4