Ammonium hydroxide, 5.0N Standardized Solution
Ammonium hydroxide, 5.0N Standardized Solution
Ammonium hydroxide, 5.0N Standardized Solution
Ammonium hydroxide, 5.0N Standardized Solution
Thermo Scientific Chemicals
Ammonium hydroxide, 5.0N Standardized Solution
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Catalog NumberQuantity
ALF035577.K21 L
Catalog number ALF035577.K2
Price (MYR)
743.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
743.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium hydroxide
CAS1336-21-6
ColorColorless
Health Hazard 1H314-H335
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c