Acetic acid, 1.0N Standardized Solution
Acetic acid, 1.0N Standardized Solution
Acetic acid, 1.0N Standardized Solution
Acetic acid, 1.0N Standardized Solution
Thermo Scientific Chemicals
Acetic acid, 1.0N Standardized Solution
CAS: 64-19-7 | C2H4O2 | 60.05 g/mol
Have Questions?
Catalog NumberQuantity
ALF035566.K21 L
Catalog number ALF035566.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetic acid
CAS64-19-7
Recommended StorageAmbient temperatures
EINECS Number200-580-7
TSCAYes
View more
Acetic acid, 1.0N standardized solution is used for volumetric analysis in laboratories.