Multi Ion anion IC standard solution, Specpure™ (Dionex) in H{2}O
Have Questions?
Catalog Number
Quantity
ALF035565.AE
100 mL
Catalog number ALF035565.AE
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialMulti Ion anion IC standard solution in H2O
Recommended StorageAmbient temperatures
OdorOdorless
EINECS Number231-791-2
TSCAYes