Fluoride, Ion chromatography standard solution, Specpure™ F∣- 1000μg/mL
Fluoride, Ion chromatography standard solution, Specpure™ F∣- 1000μg/mL
Fluoride, Ion chromatography standard solution, Specpure™ F∣- 1000μg/mL
Fluoride, Ion chromatography standard solution, Specpure™ F∣- 1000μg/mL
Thermo Scientific Chemicals
Fluoride, Ion chromatography standard solution, Specpure™ F∣- 1000μg/mL
CAS: 7681-49-4 | FNa | 41.99 g/mol
Have Questions?
Catalog NumberQuantity
ALF035550.AE100 mL
Catalog number ALF035550.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialFluoride
CAS7681-49-4
CAS7732-18-5
Recommended StorageAmbient temperatures
TSCAYes
View more
Fluoride, Ion chromatography standard solution, Specpure™, F∣- 1000μg/ml is used as a standard solution in analytical chemistry and in ion chromatography. It is used for calibration of ion chromatography and other analytical techniques.