Sulfate, Ion chromatography standard solution, Specpure™, SO4-2 1000μg/mL
Sulfate, Ion chromatography standard solution, Specpure™, SO4-2 1000μg/mL
Thermo Scientific Chemicals
Sulfate, Ion chromatography standard solution, Specpure™, SO4-2 1000μg/mL
Have Questions?
Catalog NumberQuantity
ALF035547.AE100 mL
Catalog number ALF035547.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialSulfate Ion chromatography standard solution
Name NoteSO4-2 1000μg/mL
Recommended StorageAmbient temperatures
TSCAYes
Physical FormLiquid
View more
Sulfate, Ion chromatography standard solution is used as a standard solution in analytical chemistry and in ion chromatography. It is also used for calibration of ion chromatography and other analytical techniques.