Arsenic, AAS standard solution, Specpure™ As 1000μg/mL
Arsenic, AAS standard solution, Specpure™ As 1000μg/mL
Thermo Scientific Chemicals
Arsenic, AAS standard solution, Specpure™ As 1000μg/mL
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALF033559.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialArsenic AAS standard solution
Name NoteAs 1000μg/mL
Health Hazard 1H290-H314-H335-H350
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
Health Hazard 3P201-P202-P234-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c