Potassium hydrogen phosphate trihydrate, Reagent Grade
Potassium hydrogen phosphate trihydrate, Reagent Grade
Potassium hydrogen phosphate trihydrate, Reagent Grade
Potassium hydrogen phosphate trihydrate, Reagent Grade
Thermo Scientific Chemicals
Potassium hydrogen phosphate trihydrate, Reagent Grade
CAS: 16788-57-1 | H7K2O7P | 228.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number ALF033365.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium hydrogen phosphate trihydrate
CAS16788-57-1
Recommended StorageAmbient temperatures; Store under Nitrogen
Physical FormCrystalline
Quantity1 kg
View more
Potassium hydrogen phosphate trihydrate is widely used to prepare buffer solutions in molecular biology, biochemistry and chromatography. It is involved in the production of trypticase soy agar and then utilized for culturing bacteria. As a food additive, it is used in dry powder beverages, mineral supplements and imitation dairy creamers. It is also used as a stabilizer for non-dairy coffee creamers. Further, it is a common source of phosphorous and potassium.