Potassium cyanate, 97%
Potassium cyanate, 97%
Potassium cyanate, 97%
Potassium cyanate, 97%
Thermo Scientific Chemicals
Potassium cyanate, 97%
CAS: 590-28-3 | CKNO | 81.12 g/mol
Have Questions?
Catalog NumberQuantity
ALF033356.22100 g
Catalog number ALF033356.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium cyanate
Melting Point700°C to 900°C (decomposition)
CAS590-28-3
ColorWhite
Health Hazard 1H302
View more
Potassium cyanate is used in gold mining, organic synthesis and electroplating.