Potassium hydrogen phthalate, primary standard, ACS, 99.95-100.05%
Potassium hydrogen phthalate, primary standard, ACS, 99.95-100.05%
Potassium hydrogen phthalate, primary standard, ACS, 99.95-100.05%
Potassium hydrogen phthalate, primary standard, ACS, 99.95-100.05%
Thermo Scientific Chemicals
Potassium hydrogen phthalate, primary standard, ACS, 99.95-100.05%
CAS: 877-24-7 | C8H5KO4 | 204.222 g/mol
Have Questions?
Catalog NumberQuantity
ALF033351.30250 g
Catalog number ALF033351.30
Price (MYR)
500.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
500.00
EA
Add to cart
Specifications
Chemical Name or MaterialPotassium hydrogen phthalate, primary standard
Melting Point297°C (decomposition)
Name NoteFor ACS analysis
CAS877-24-7
ColorWhite
View more
As a primary standard for preparing volumetric alkali solutions and as a buffer in pH determinations. Potassium hydrogen phthalate is used as a primary standard for acid- base titrations as well as for calibrating pH meters. It acts as a buffer in pH determination. It finds application as a useful standard for total organic carbon testing.