Hydrogen peroxide, ACS, 29-32% w/w aq. soln., stab.
Hydrogen peroxide, ACS, 29-32% w/w aq. soln., stab.
Hydrogen peroxide, ACS, 29-32% w/w aq. soln., stab.
Hydrogen peroxide, ACS, 29-32% w/w aq. soln., stab.
Thermo Scientific Chemicals
Hydrogen peroxide, ACS, 29-32% w/w aq. soln., stab.
For Bleaching and Deodorizing Textiles, Wood, Hair | CAS: 7722-84-1 | H2O2 | 34.014 g/mol
Have Questions?
Catalog NumberQuantity
ALF033323.AP500 mL
Catalog number ALF033323.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialHydrogen peroxide
ColorColorless
CAS7722-84-1
Boiling Point107°C
Health Hazard 1H271-H302+H332-H314
View more
For bleaching and deodorizing textiles, wood, hair, etc; rocket propulsion; maturing agent in food; water and sewage treatment; disinfectant; laboratory reagentHydrogen peroxide is used as an oxidizing agent to prepare propylene oxide, dibenzoyl peroxide, lauryl peroxide, peracetic acid and meta-chloroperoxybenzoic acid. It is also involved in the production of inorganic chemicals like sodium perborate and sodium percarbonate, which are used as bleaching agents in detergents. It is an associated iron catalyst and utilized for waste-water treatment to remove organic impurities through an advanced oxidation process. It is also used as a laboratory disinfectant and as a topical antiseptic. In the cosmetic industry, it is used for bleaching human hair and tooth whitening by mixing with ammonium hydroxide and baking soda respectively. Monopropellant engines using hydrogen peroxide as the fuel are commonly used for air-independent underwater propulsion applications.