tert-Butyl alcohol, ACS, 99+%
tert-Butyl alcohol, ACS, 99+%
tert-Butyl alcohol, ACS, 99+%
tert-Butyl alcohol, ACS, 99+%
Thermo Scientific Chemicals
tert-Butyl alcohol, ACS, 99+%
CAS: 75-65-0 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Catalog number ALF033278.K2
Price (MYR)
544.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
544.00
EA
Add to cart
Specifications
Chemical Name or MaterialTert-Butyl alcohol
Name NoteFor ACS analysis
CAS75-65-0
Health Hazard 1H225-H319-H332-H335-H336
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
View more
tert-Butyl alcohol is used as a solvent and an ethanol denaturant. It is an active ingredient in paint remover. It acts as an intermediate in the production of methyl tertiary butyl ether and ethyl tertiary butyl ether by reaction with methanol and ethanol. It is also used in the preparation of tert-butyl chloride, tert-butyl hydroperoxide, isobutylene and methyl methacrylate. It serves as a fuel additive as well as a gasoline octane booster.