Succinic acid, ACS, 99.0% min
Succinic acid, ACS, 99.0% min
Succinic acid, ACS, 99.0% min
Succinic acid, ACS, 99.0% min
Thermo Scientific Chemicals
Succinic acid, ACS, 99.0% min
CAS: 110-15-6 | C4H6O4 | 118.09 g/mol
Have Questions?
Catalog NumberQuantity
ALF033272.36500 g
Catalog number ALF033272.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSuccinic acid
CAS110-15-6
Melting Point186°C to 188°C
Boiling Point235°C
Health Hazard 1H318-H335
View more
Succinic acid acts as a leaching agent in extracting lithium, cobalt from used Li-ion batteries and magnesium from magnesite ore. It is used as a starting material in the synthesis of new elastic polyesters, fumaric acid, succinic anhydride and alkyd resins. It plays an important role as an acidity regulator in the food and beverage industry; as an excipient in pharmaceutical products and acts as a cocrystallising agent in organic synthesis. Its derivative viz diethyl ester is used as a substrate in the Stobbe condensation.