Silicic acid hydrate
Silicic acid hydrate
Silicic acid hydrate
Silicic acid hydrate
Thermo Scientific Chemicals
Silicic acid hydrate
CAS: 1343-98-2 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF033270.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
AppearanceWhite powder
Chemical Name or MaterialSilicic Acid Hydrate
ColorWhite
CAS1343-98-2
Health Hazard 1H373: May cause damage to organs through prolonged or repeated exposure.
View more
Silicic acid hydrate is used for studying neurodegenerative diseases due to its ability to inhibit aluminum uptake. It is utilized for the manufacture catalyst and catalyst carrier. It is involved in tungsten filament production as chemical reagent and flux. Further, it is used for oil and wax decoloring.