Amidosulfonic acid, ACS, 99.3-100.3% (Assay dried basis)
Amidosulfonic acid, ACS, 99.3-100.3% (Assay dried basis)
Amidosulfonic acid, ACS, 99.3-100.3% (Assay dried basis)
Amidosulfonic acid, ACS, 99.3-100.3% (Assay dried basis)
Thermo Scientific Chemicals
Amidosulfonic acid, ACS, 99.3-100.3% (Assay dried basis)
CAS: 5329-14-6 | H3NO3S | 97.088 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF033233.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmidosulfonic acid
pH1.2
CAS5329-14-6
Melting Point∼205°C (decomposition)
Health Hazard 1H290-H315-H319
View more
Amidosulfonic acid is widely utilized as a green catalyst for the preparation of amide from ketoxime. It finds an application as a titrant in the determination of the burette injection volume and chemical calibration factor. It is an efficient catalyst in the synthesis of polyhydroquinoline derivatives by Hantzsch condensation reaction, deazaoxaflavin, fatty acid methyl and ethyl esters and in the isolation of 5-hydroxymethylfurfural (HMF) from bamboo fiber. It is involved in the determination of silicates in water samples. It acts as a neutralizing agent during the determination of paracetamol by Glynn and Kendal colorimetric method.