Manganese(II) iodide, anhydrous, 98%
Manganese(II) iodide, anhydrous, 98%
Manganese(II) iodide, anhydrous, 98%
Manganese(II) iodide, anhydrous, 98%
Thermo Scientific Chemicals
Manganese(II) iodide, anhydrous, 98%
CAS: 7790-33-2
Have Questions?
Catalog NumberQuantity
ALF032808.0910 g
Catalog number ALF032808.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialManganese(II) iodide
Melting Point638°C
Name NoteAnhydrous
CAS7790-33-2
ColorRed to White
View more
Manganese(II) iodide can be used as a pink pigment or as a source of the manganese ion or iodide ion. It is often used in the lighting industry.