Bromophenol Blue, ACS
Bromophenol Blue, ACS
Bromophenol Blue, ACS
Bromophenol Blue, ACS
Thermo Scientific Chemicals
Bromophenol Blue, ACS
CAS: 115-39-9 | C19H10Br4O5S | 669.96 g/mol
Have Questions?
Catalog NumberQuantity
ALF032641.1425 g
Catalog number ALF032641.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBromophenol Blue
Melting Point270°C (decomposition)
CAS115-39-9
Health Hazard 1H500
Recommended StorageAmbient temperatures
View more
Bromophenol blue is used as a tracking dye in electrophoresis, as a color marker in gel electrophoresis and polyacrylamide gel electrophoresis. As a biological stain, it plays an important role to stain proteins and nucleic acids. It acts an intermediate and an acid-base indicator in the pH range 3.0 to 4.6.