Bromothymol Blue, ACS
Bromothymol Blue, ACS
Bromothymol Blue, ACS
Bromothymol Blue, ACS
Thermo Scientific Chemicals
Bromothymol Blue, ACS
CAS: 76-59-5 | C27H28Br2O5S | 624.384 g/mol
Have Questions?
Catalog NumberQuantity
ALF032638.065 g
Catalog number ALF032638.06
Price (MYR)
279.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
279.00
EA
Add to cart
Specifications
Chemical Name or MaterialBromothymol Blue
Melting Point200°C to 202°C
CAS76-59-5
Recommended StorageAmbient temperatures
RTECS NumberSJ450000
View more
Bromothymol Blue is a pH indicator used for measuring pH around 7 in pools and fish tanks. It operates in the pH range 6.0 to 7.6 due to the presence of an electron withdrawing and donating groups like bromine atoms and alkyl substitutents respectively. In the laboratory, it is used as a biological slide stain as well as used in obstetrics for detecting premature rupture of membranes. Further, it is used for the observation of photosynthetic activities and a respiratory indicator. In addition to this, it is used to define cell walls or nuclei under the microscope.