Chloroform, ACS, 99.8+%
Chloroform, ACS, 99.8+%
Chloroform, ACS, 99.8+%
Chloroform, ACS, 99.8+%
Thermo Scientific Chemicals
Chloroform, ACS, 99.8+%
CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Have Questions?
Catalog NumberQuantity
ALF032614.K21 L
Catalog number ALF032614.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialChloroform
Melting Point-63°C
CAS67-66-3
Health Hazard 1H302-H315-H319-H331-H335-H336-H351-H361d-H373
Health Hazard 3P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
View more
Chloroform is suitable for pesticide analysis, environmental and GC analysis. It is a commonly used laboratory solvent. It was previously used as an anesthetic. It is also used as a precursor to PTFE and refrigerants. Chloroform was used in the past as an extraction solvent for fats, oils and greases. It is useful for the rapid and simple release of periplasmic proteins. The mixture of guanidinium thiocyanate-phenol-chloroform plays an essential role in the RNA isolation process.