1-Butanol, Ultrapure, Spectrophotometric Grade, 99.0+%
1-Butanol, Ultrapure, Spectrophotometric Grade, 99.0+%
1-Butanol, Ultrapure, Spectrophotometric Grade, 99.0+%
1-Butanol, Ultrapure, Spectrophotometric Grade, 99.0+%
Thermo Scientific Chemicals
1-Butanol, Ultrapure, Spectrophotometric Grade, 99.0+%
CAS: 71-36-3 | C4H10O | 74.123 g/mol
Have Questions?
Catalog NumberQuantity
ALF032443.K21 L
Catalog number ALF032443.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or Material1-Butanol
pH7
Melting Point-90°C
CAS71-36-3
ColorColorless
View more
1-Butanol is primarily used as a solvent, as an intermediate in chemical synthesis, and as a fuel. It is also used as a paint thinner and a solvent in other coating applications where it is used as a relatively slow evaporating latent solvent in lacquers and ambient-cured enamels. It finds other uses such as a component of hydraulic and brake fluids.