Dimethyl sulfoxide, Spectrophotometric Grade, 99.9+%
Dimethyl sulfoxide, Spectrophotometric Grade, 99.9+%
Dimethyl sulfoxide, Spectrophotometric Grade, 99.9+%
Dimethyl sulfoxide, Spectrophotometric Grade, 99.9+%
Thermo Scientific Chemicals
Dimethyl sulfoxide, Spectrophotometric Grade, 99.9+%
CAS: 67-68-5 | C2H6OS | 78.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number ALF032434.K7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Specifications
Chemical Name or MaterialDimethyl sulfoxide
Melting Point18.4°C
CAS67-68-5
ColorColorless
Health Hazard 1H227
View more
Dimethyl sulfoxide is used in spectrophotometry and environmental testing.