Mineral oil
Thermo Scientific Chemicals
Mineral oil
CAS: 8020-83-5
Have Questions?
Catalog NumberQuantity
ALF031911.A11 kg
Catalog number ALF031911.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialMineral oil
CAS8020-83-5
Recommended StorageAmbient temperatures
TSCANo
Flash Point>100°C (212°F)
View more
Mineral oil is used as an overlay covering microdrops of culture medium in petri dishes. It is an active ingredient in baby lotions, cold creams, ointments and cosmetics. It is also used in electrical and mechanical industries as thermal fluid in electrical components and non -conductive coolant. It finds application as a transformer oil, as an anti rust agent for blades and as an insulator in high voltage switchgear. Further, it is used to store reactive metals like lithium, sodium, etc and in X-ray crystallography to suspend crystals.