n-Undecane, 99%
n-Undecane, 99%
n-Undecane, 99%
Thermo Scientific Chemicals
n-Undecane, 99%
CAS: 1120-21-4 | C11H24 | 156.31 g/mol
Have Questions?
Catalog NumberQuantity
ALF031807.1850 g
Catalog number ALF031807.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Materialn-Undecane
CAS1120-21-4
Health Hazard 1H227-H304
Health Hazard 2GHS H Statement
H304-H226-H315-H319-H335
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P280-P301+P310-P331-P370+P378q-P501c
View more
n-Undecane is used as a mild sex attractant for various types of moths and cockroaches. It is also used as an internal standard in the gas chromatographic analysis. Further, it is used as lubricants and lubricant additives.