n-Hexatriacontane, 97+%
n-Hexatriacontane, 97+%
n-Hexatriacontane, 97+%
n-Hexatriacontane, 97+%
Thermo Scientific Chemicals
n-Hexatriacontane, 97+%
CAS: 630-06-8 | C36H74 | 506.99 g/mol
Have Questions?
Catalog NumberQuantity
ALF031641.0910 g
Catalog number ALF031641.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or Materialn-Hexatriacontane
CAS630-06-8
Melting Point76.2°C
ColorWhite
Recommended StorageAmbient temperatures
View more
n-Hexatriacontane is used as a paraffin wax and in candles. It is used as a key starting material for the preparation of lower alkanes by cracking. It is also used for example in road surfacing.