Nickel on silica-alumina, catalyst
Nickel on silica-alumina, catalyst
Nickel on silica-alumina, catalyst
Nickel on silica-alumina, catalyst
Thermo Scientific Chemicals
Nickel on silica-alumina, catalyst
Formula: 66±5% Ni | CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF031276.22
Price (MYR)
706.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
706.00
EA
Add to cart
Specifications
Chemical Name or MaterialNickel
ColorBlack
Name Noteon silica-alumina, catalyst
CAS7440-02-0
Health Hazard 1H228-H317-H350i-H372
View more
Nickel on Silica-Alumina Catalys products are often used as the most efficient way to catalyze organic reactions. It is also used in R&D, manufacture of chemicals, Pharmaceuticals and intermediates. It is an important raw material and intermediate used in Organic Synthesis.