1,2-Propanediol, ACS, 99.5% min
1,2-Propanediol, ACS, 99.5% min
1,2-Propanediol, ACS, 99.5% min
1,2-Propanediol, ACS, 99.5% min
Thermo Scientific Chemicals
1,2-Propanediol, ACS, 99.5% min
As a substitute for ethylene glycol and glycerol. Good solvent for resins. Dissolves many essential oils, but immiscible with fixed oils. | CAS: 57-55-6 | C3H8O2 | 76.095 g/mol
Have Questions?
Catalog NumberQuantity
ALF030948.K21 L
Catalog number ALF030948.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or Material1,2-Propanediol
CAS57-55-6
Melting Point-60°C
Recommended StorageAmbient temperatures; Store under Nitrogen
Density1.036
View more
1,2-Propanediol, ACS is used as a substitute for ethylene glycol and glycerol. It is a good solvent for resins. It dissolves many essential oils but is immiscible with fixed oils.