n-Tetradecane, 99+%
n-Tetradecane, 99+%
n-Tetradecane, 99+%
Thermo Scientific Chemicals
n-Tetradecane, 99+%
CAS: 629-59-4 | C14H30 | 198.394 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF030809.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Materialn-Tetradecane
CAS629-59-4
Health Hazard 1H304
Health Hazard 3P301+P310-P331-P501c
Melting Point6°C
View more
n-Tetradecane is used to study the thermodiffusion behavior of the microemulsion droplets of water, n-alkanes and pentaethylene glycol monododecyl ether. It is used as reference material in gas chromatographic analysis, as a solvent and finds applications in organic synthesis.