Salicylic acid, ACS, 99+%
Salicylic acid, ACS, 99+%
Salicylic acid, ACS, 99+%
Salicylic acid, ACS, 99+%
Thermo Scientific Chemicals
Salicylic acid, ACS, 99+%
CAS: 69-72-7 | C7H6O3 | 138.12 g/mol
Have Questions?
Catalog NumberQuantity
ALF030782.22100 g
Catalog number ALF030782.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSalicylic acid
CAS69-72-7
Melting Point158°C to 160°C
Boiling Point211°C (20mmHg)
Health Hazard 1H302-H318-H361d
View more
Salicylic acid is used as a food preservative as well as used in the treatment of many skin disorders such as acne, dandruff, psoriasis and seborrheic dermatitis of the skin and scalp. It is an antiseptic. It is mainly used in the pharmaceutical industry for the manufacture of antipyretic, analgesic, anti-inflammatory and diuretic drugs. It is also used in the dye industry for making azo direct dyes and acidic mordant dyeing.