Thioacetamide, ACS, 99% min
Thioacetamide, ACS, 99% min
Thioacetamide, ACS, 99% min
Thioacetamide, ACS, 99% min
Thermo Scientific Chemicals
Thioacetamide, ACS, 99% min
CAS: 62-55-5 | C2H5NS | 75.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALF030638.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialThioacetamide
CAS62-55-5
Health Hazard 1H302-H315-H319-H336-H351-H373
Health Hazard 2GHS H Statement
H350-H302-H315-H319
May cause cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
View more
Thioacetamide is used in qualitative inorganic analysis as an in-situ source for sulfide ions. It is used in analytical chemistry as a source of hydrogen sulfide. It is an additive in enantioselective reduction of beta-keto esters with immobilized bakers yeast. It is also used as a vulcanizing agent of polymer and a crosslinking agent.