Nitrite, Quant™ Test Strips
Nitrite, Quant™ Test Strips
Nitrite, Quant™ Test Strips
Have Questions?
Catalog NumberQuantity
ALF023283.QA1 Kit
Catalog number ALF023283.QA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 Kit
Request bulk or custom format
Specifications
Quantity1 Kit
RangeMeasures 1 to 50ppm NO2- in aqueous solutio
TypeNitrite test strip