Water, ultrapure, HPLC Grade
Water, ultrapure, HPLC Grade
Water, ultrapure, HPLC Grade
Water, ultrapure, HPLC Grade
Thermo Scientific Chemicals
Water, ultrapure, HPLC Grade
CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number ALF022934.K7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Specifications
Chemical Name or MaterialWater
CAS7732-18-5
Melting Point0°C
Boiling Point100°C
Recommended StorageAmbient temperatures
View more
Water ultrapure is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry. It is also used in conductometric titration.