N,N-Dimethylacetamide, HPLC Grade, 99.5+%
N,N-Dimethylacetamide, HPLC Grade, 99.5+%
N,N-Dimethylacetamide, HPLC Grade, 99.5+%
N,N-Dimethylacetamide, HPLC Grade, 99.5+%
Thermo Scientific Chemicals
N,N-Dimethylacetamide, HPLC Grade, 99.5+%
CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Catalog NumberQuantity
ALF022916.K21 L
Catalog number ALF022916.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialN, N-Dimethylacetamide
Melting Point-20°C
CAS127-19-5
ColorColorless
Health Hazard 1H227-H312+H332-H360D
View more
N,N-Dimethylacetamide is used in all analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS).