N,N-Dimethylformamide, HPLC Grade, 99.7+%
N,N-Dimethylformamide, HPLC Grade, 99.7+%
N,N-Dimethylformamide, HPLC Grade, 99.7+%
N,N-Dimethylformamide, HPLC Grade, 99.7+%
Thermo Scientific Chemicals
N,N-Dimethylformamide, HPLC Grade, 99.7+%
CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Catalog NumberQuantity
ALF022915.K21 L
Catalog number ALF022915.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialN, N-Dimethylformamide
Melting Point61°C
CAS68-12-2
ColorColorless
Health Hazard 1H226-H312+H332-H319-H335-H336-H350-H360D
View more
N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films. It is used as a mobile phase in High Performance Liquid Chromatography.