Dimethyl sulfoxide, HPLC grade, >99.9%, Thermo Scientific™
Dimethyl sulfoxide, HPLC grade, >99.9%, Thermo Scientific™
Dimethyl sulfoxide, HPLC grade, >99.9%, Thermo Scientific™
Dimethyl sulfoxide, HPLC grade, >99.9%, Thermo Scientific™
Thermo Scientific Chemicals
Dimethyl sulfoxide, HPLC grade, >99.9%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALF022914.0F2500 mL
Catalog number ALF022914.0F
Price (MYR)
1,589.00
EA
Add to cart
Quantity:
2500 mL
Request bulk or custom format
Price (MYR)
1,589.00
EA
Add to cart
Specifications
Chemical Name or MaterialDimethyl sulfoxide
Melting Point18.4°C
CAS67-68-5
ColorColorless
Health Hazard 1H227
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.