Pyridine, HPLC Grade, 99.5+%
Pyridine, HPLC Grade, 99.5+%
Pyridine, HPLC Grade, 99.5+%
Pyridine, HPLC Grade, 99.5+%
Thermo Scientific Chemicals
Pyridine, HPLC Grade, 99.5+%
Maximum level of impurities: Evaporation residue 4ppm, H2O 0.05% | CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Catalog NumberQuantity
ALF022905.K21 L
Catalog number ALF022905.K2
Price (MYR)
1,233.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
1,233.00
EA
Add to cart
Specifications
Chemical Name or MaterialPyridine
Melting Point-42°C
CAS110-86-1
Health Hazard 1H225-H302+H312+H332-H315-H319
Health Hazard 3P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
View more
Pyridine is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry