Toluene, HPLC Grade, 99.7% min
Toluene, HPLC Grade, 99.7% min
Toluene, HPLC Grade, 99.7% min
Toluene, HPLC Grade, 99.7% min
Thermo Scientific Chemicals
Toluene, HPLC Grade, 99.7% min
CAS: 108-88-3 | C7H8 | 92.14 g/mol
Have Questions?
Catalog NumberQuantity
ALF022903.K21 L
Catalog number ALF022903.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialToluene
Melting Point93°C
CAS108-88-3
Health Hazard 1H225-H304-H315-H319-H336-H361d-H373
Health Hazard 3P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
View more
Toluene is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.