2,2,4-Trimethylpentane, HPLC Grade, 99.7+%
2,2,4-Trimethylpentane, HPLC Grade, 99.7+%
2,2,4-Trimethylpentane, HPLC Grade, 99.7+%
2,2,4-Trimethylpentane, HPLC Grade, 99.7+%
Thermo Scientific Chemicals
2,2,4-Trimethylpentane, HPLC Grade, 99.7+%
Maximum level of impurities: Evaporation residue 3ppm, Chlorinated pesticides 5ppt, H2O 0.02% | CAS: 540-84-1 | C8H18 | 114.232 g/mol
Have Questions?
Catalog NumberQuantity
ALF022901.K21 L
Catalog number ALF022901.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or Material2, 2, 4-Trimethylpentane
CAS540-84-1
Health Hazard 1H225-H304-H315-H319-H336
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c
Melting Point°C to 107°C
View more
2,2,4-Trimethylpentane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.