Mullite
Mullite
Mullite
Thermo Scientific Chemicals
Mullite
CAS: 1302-93-8 | Al6O13Si2 | 426.05 g/mol
Have Questions?
Catalog NumberQuantity
ALF022881.36500 g
Catalog number ALF022881.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMullite
CAS1302-93-8
ColorWhite
Recommended StorageAmbient temperatures
Physical FormPowder
View more
Mullite is present in the form of needles in porcelain, it is used in high-mullite based products is in hot blast stove checker bricks, melting and firing processes, and is used as a refractory material, manufacture of a type of crucible, steel industry, electronic substrates and protective coatings.